Learn More
FUJIFILM IRVINE SCIENTIFIC INC IS Sf Insect, 1 L
Supplier: FUJIFILM IRVINE SCIENTIFIC INC 911741L
IS Sf Insect delivers reliable growth of Sf9 and Sf21 insect cells in suspension cultures, and supports Baculovirus Expression Vector Systems (BEVS) for robust protein expression, including recombinant proteins and viral-like particles, as well as viral vector production.