Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Iron-Supplemented Calf Serum
Iron-Supplemented Calf Serum (ISCS) is supplemented with ferric salt, enabling this serum to bind from three to four times the iron of Fetal Bovine Serum.
Supplier: Novus Biologicals™ S11995NOV
Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Bovine | |
50 mL |
Iron-Supplemented Calf Serum | |
Calf Bovine Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title