Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Iron-Supplemented Calf Serum, Heat Inactivated
Iron-Supplemented Calf Serum (ISCS) is supplemented with ferric salt, enabling this serum to bind from three to four times the iron of Fetal Bovine Serum.
Supplier: Novus Biologicals™ S11950HNOV
Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Bovine | |
500 mL |
Iron-Supplemented Calf Serum, Heat Inactivated | |
Calf Bovine Serum |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title