Learn More
Newcomer Supply Iron Control Slides, 98/set
Supplier: Newcomer Supply 4320B
Newcomer Supply Iron Control Slides are for the positive histochemical staining of ferric iron deposits in tissue sections. Iron Control Slides contain sections of positive staining liver or spleen.