Learn More
OriGene IRAKM (IRAK3) (NM_007199) Human Tagged ORF Clone | 10ug
Supplier: OriGene RG209245
This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene. The ORF clone is ion-exchange column purified, transfection-ready dried plasmid DNA, and shipped with 2 vector sequencing primers.