Learn More
Omega Scientific Inc Thermometer Infrared, Fda Cert
SDP
Supplier:  Omega Scientific Inc IDT300
Encompass_Preferred
IDT-300: Non-Contact Infrared Digital ThermometerNon Contact, 1 Second Measurement Infrared Digital Thermometer with LCD Display One second measurement with Non-contact infrared.Measuring distance is 1″ – 2″.Auto shutdown feature for power saving.Measure surface temperatures ranging from 89.6℉ to 109.22 ℉ (32℃ to 42℃)Easily switch between Fahrenheit and Celsius.Operating Voltage DC 3VMemory Recall (32 Measurements)Material: Plastic
Catalog No. NC1854657
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.