Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ IonSense™ PDMS/DVB SPE-it Tips
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supplier: MilliporeSigma™ Supelco™ 57249U
Description
Low cost, micro-sample prep devices consisting of a coated microfiber held in place in a micropipette tip making it compatible with both multi-channel pipettors and robotic liquid handling systems. The tips are sold in a 96 well tray configuration.Specifications
SPE | |
Food and Beverages | |
IonSense | |
96 Tips/Pk. |
Fiber L: 1 cm | |
Pipettor or Robotic Liquid Handling Systems | |
SPE Tip |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
IonSense is a registered trademark of IonSense, Inc.