Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ IonSense™ C18 SPE-it Tips
Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.
Supplier: MilliporeSigma™ Supelco™ 57264U
Description
IonSense C18 SPE-it Tips are solid phase extraction devices configured to permit isolation of analytes from aqueous solutions and then direct analysis without further sample processing.Specifications
SPE | |
Food and Beverages | |
IonSense | |
96 Tips/Pk. |
Fiber L: 1 cm | |
Pipettor or Robotic Liquid Handling Systems | |
SPE Tip |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
IonSense is a registered trademark of IonSense, Inc.