Learn More
Thermo Scientific ION PACK AS 4 A 4MM COLUMN
Supplier: Thermo Scientific 037041
IONPAC AS4A Analytical Column, 4 x 250mm, The IonPac AS4A should operate at a backpressure between 700 to 1000 psi at 2mL/min
This item is not returnable.
View return policy