Learn More
Thermo Scientific Dionex™ IonPac™ CS19 IC Columns
Supplier: Thermo Scientific 076028
Dionex™ IonPac™ CS19 IC Columns, 2mm x 250mm. This high-capacity, carboxylate-functionalized column is specifically optimized for the separation of common cations, small polar amines, and moderately hydrophobic and polyvalent cations using simple aqueous eluents and elevated temperatures. The column is ideal for analysis of power plant waters treated with ammonium, morpholine, or ethanolamine and can also be used to analyze chemical additives, chemical process solutions, scrubber solutions, personal care products, and food samples.