Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
Description
Honeywell ACS solvents for general laboratory use are:
- a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested tmeet ACS general use specifications
Specifications
Physical Form | Solid |
Quantity | 5 g |
UN Number | 3495 |
Beilstein | 3587194 |
IUPAC Name | diiodine |
Formula Weight | 253.81g/mol |
Percent Purity | ≥99.8% |
Grade | ACS Reagent |
Vapor Pressure | 0.31mmHg (25°C), 1mmHg (38.7°C) |
Density | Vapor Density: 9 (vs air) |
Show More |
Safety and Handling
Hazard Statement | P261-P273-P280-P305 + P351 + P338-P314 |
Precautionary Statement | H312 + H332-H315-H319-H335-H372-H400 |
EINECSNumber | 231-442-4 |
RTECSNumber | NN1575000 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction