Learn More
Abbott™ Inverness Cardiac Risk Assessment Kits
Supplier: Abbott™ 339100
This item is not returnable.
View return policy
Specifications
High | |
Macra Lp(a) ELISA Test Kit | |
12 x 8-well strips with breakaway wells, sample diluent, control, calibrators, conjugate, substrate, Wash Buffer, stop solution |
Serum or Plasma | |
Helps to quantitatively measure an important cardiac risk marker lipoprotein in Serum or Plasma | |
100 Pack |