Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alere Inverness Clearview™ H. pylori II Test Kit
Patented technology provides highly sensitive and specific detection of H. pylori in human serum, plasma, or whole blood
Supplier: Alere 6706KCV
Includes: 30 individually pouched test devices, each containing one disposable transfer pipette, a developer buffer, positive and negative controls, two directional inserts (one for waived and one for non-waived) and one procedure card
Description
CLIA Complexity: Waived for whole blood; Moderate for serum or plasma
- Sensitivity: 92.6%
- Specificity: 82.4%
- Easy two-step procedure
- Built-in control ensures accuracy
- Use with multiple sample tyes - whole blood, serum, or plasma
- Positive results in as soon as 1 minute; negative results, less than 5 minutes
- Room temperature storage with 18 months shelf life
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction