Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ Polypropylene Beard Net
Click to view available options
Beard Net, white
Description
Disposable Beard Restraint and Cover
Specifications
Type | General Purpose Beard Cover |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Latex Free | Yes |
For Use With (Application) | Used to protect against dirt, grime and certain dry particulates in non-hazardous environments |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction