Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Internal Standard, Triphenyl Phosphate, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
115-86-6, 1634-04-4
C18H15O4P
326.29
MFCD00003031
XZZNDPSIHUTMOC-UHFFFAOYSA-N
8289
CHEBI:35033
triphenyl phosphate
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
115-86-6, 1634-04-4
C18H15O4P
326.29
MFCD00003031
XZZNDPSIHUTMOC-UHFFFAOYSA-N
8289
CHEBI:35033
triphenyl phosphate
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Triphenyl phosphate115-86-6500μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.05, 99.95
CAS Max % 0.05, 99.95
Packaging 2mL amber prescored glass ampule
Methods For US EPA Method 507
Solvent or Matrix MTBE
Quantity 1mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Triphenyl Phosphate, USEPA Method 507

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only