Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Internal Standard, Pentachloronitrobenzene, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
82-68-8, 1634-04-4
C6Cl5NO2
295.321
LKPLKUMXSAEKID-UHFFFAOYSA-N
6720
CHEBI:34908
1,2,3,4,5-pentachloro-6-nitrobenzene
C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]
82-68-8, 1634-04-4
C6Cl5NO2
295.321
LKPLKUMXSAEKID-UHFFFAOYSA-N
6720
CHEBI:34908
1,2,3,4,5-pentachloro-6-nitrobenzene
C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)[N+](=O)[O-]

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Pentachloronitrobenzene82-68-8100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.01, 99.99
CAS Max % 0.01, 99.99
Packaging 2mL amber prescored glass ampule
Methods For US EPA Method 508
Solvent or Matrix MTBE
Quantity 1mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Pentachloronitrobenzene, USEPA Method 508

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only