Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Internal Standard, 4-Bromo-3,5-dimethylphenyl-n-methylcarbamate, SPEX CertiPrep™

Click to view available options
Quantity:
1mL
Packaging:
2mL amber prescored glass ampule
672-99-1, 75-05-8
C10H12BrNO2
258.12
MFCD00027941
ZOZFMTULOYRWEL-UHFFFAOYSA-N
688148
(4-bromo-3,5-dimethylphenyl) N-methylcarbamate
CNC(=O)OC1=CC(C)=C(Br)C(C)=C1
672-99-1, 75-05-8
C10H12BrNO2
258.12
MFCD00027941
ZOZFMTULOYRWEL-UHFFFAOYSA-N
688148
(4-bromo-3,5-dimethylphenyl) N-methylcarbamate
CNC(=O)OC1=CC(C)=C(Br)C(C)=C1

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
4-Bromo-3,5-dimethylphenyl-N-methylcarbamate672-99-1100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.01, 99.99
CAS Max % 0.01, 99.99
Packaging 2mL amber prescored glass ampule
Methods For US EPA Methods 531, 531.1
Solvent or Matrix Acetonitrile
Quantity 1mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material 4-Bromo-3,5-dimethylphenyl-n-methylcarbamate, USEPA Method 531.1

DOTInformation : Nonhazardous

Recommended Storage : Ambient

Product intended for laboratory use only