Learn More
Pendotech Pendotech Pressuremat To Watson Marlow Interface Cable
Supplier:  Pendotech PMATCBWM15
Encompass_Preferred
Interface cable for Watson Marlow 120U/DV pump with D15 connector to PMAT to start/stop function, 6 feet (2meters)(Also works on the 101 model)
Catalog No. NC1656831