Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Ribbon-Type Iris Scissors

$409.00 - $474.00

Specifications

Product Type Iris Ribbon Scissors
Blade Style Straight
Grade Premium
Material Stainless Steel
For Use With (Application) Surgical, ophthalmic
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Price Quantity & Availability  
12-460-272
View Documents
Integra™ Miltex™
181415
4 in. 10.16 cm
Each for $409.00
Add to Cart
 
12-460-273
View Documents
Integra™ Miltex™
181417
3.7 in. 9.3 cm
Each for $474.00
Add to Cart
 
Description

Description

  • Stainless steel
  • Ribbon type ringhandles
  • Straight

Specifications

Specifications

Iris Ribbon Scissors
Premium
Surgical, ophthalmic
Straight
Stainless Steel
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit