Learn More
Glas Col LLC Insulating Mantle
Supplier: Glas Col LLC 100A INS1023
Fabric insulating top for 250ml 1, 2 or 3 neck spherical flask.With no heat serves to reduce vapor condensation on top half of flask and to protect your glassware. For use with aluminum housed heating mantles.