Learn More
LaMotte™ Insta-Test™ Water Test Strips
Perform safe, quick and easy water quality checks with Insta-Test™ Water Test Strips.
Supplier: LaMotte™ 2997
This item is not returnable.
View return policy
Description
- Monitor water without having to use reagents or field kits.
Order Info
Each vial of test strips contains 50 tests per factor, except S66233, which contains 25 tests per factor.
Specifications
Alkalinity | |
Drinking, food/beverage, pool water testing | |
0 to 180 ppm (0, 40, 80, 120, 180 ppm) |
Strip | |
50 Pk. | |
Water Test Strip |