Learn More
Fine Science Tools INSERT PIN STRAIGHT POINT
Supplier: Fine Science Tools 2600702
Insert Pins / Tip Shape: Straight Point/ Rod Diameter: 0.6mm/ Alloy: Stainless Steel/ Length: 5cm/ Unit: Each