Learn More
Fine Science Tools Insect Pins, Tip: 0.03 mm, 4 cm, 100/pk
Supplier: Fine Science Tools 2600125
Insect Pins - Stainless Steel/Size 000/ Size: 000/ Tip Diameter: 0.03mm/ Rod Diameter: 0.25mm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: 100