Learn More
Fine Science Tools Insect Pins, Tip: 0.03 mm, 4 cm, 100/pk
Supplier: Fine Science Tools 2600040
Insect Pins - Black Anodized/Size 1/ Size: 1/ Tip Diameter: 0.03mm/ Rod Diameter: 0.4mm/ Alloy: Black Anodized Steel/ Length: 4cm/ Unit: 100