Learn More
Fine Science Tools INSECT PIN .3MM SZ00 BLK C/PK
Supplier: Fine Science Tools 2600030
Insect Pins - Black Anodized/Size 00/ Size: 00/ Tip Diameter: 0.03mm/ Rod Diameter: 0.3mm/ Alloy: Black Anodized Steel/ Length: 4cm/ Unit: 100