Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Osmosis and Diffusion Lab
SCI_ED

Allows you to learn about two forms of passive transport: diffusion and osmosis.

Supplier:  Innovating Science™ IS3001

Catalog No. S98658A

This item has been discontinued and is no longer available. Please reach out to your sales representative.


Includes:
  • Dialysis tubing
  • Starch solution (100mL of 1.0%)
  • Sucrose
  • Iodine/potassium iodide solution (15mL)
  • Plastic cups
This item is not returnable. View return policy

Description

Description

  • Compare and contrast similarities and differences in the processes of diffusion and osmosis
  • Colorimetric test demonstrates the movement of a solute across a semi-permeable membrane
  • Environment likely to facilitate osmosis
  • Data is gathered to determine whether or not osmosis may have occurred

Order Info

Materials for 15 groups.

Specifications

Specifications

Osmosis and Diffusion Lab Kit
7 to 14
Helps students to learn about two forms of passive transport: diffusion and osmosis
1
15 Lab Groups
White
13 to 20
Teacher's manual and student study guide copymasters, 30 piece dialysis tubing, starch, sucrose bottle, 15ml iodine/potassium iodide solution bottle, 30 plastic cups
9.75 x 6.25 x 4 in.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit