Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ AP Biology Investigation Kit No. 5: Photosynthesis
SCI_ED

Students learn necessary components and conditions for photosynthesis using leaf disks to measure oxygen accumulation and its affect on the process.

Supplier:  Innovating Science™ IS3705

Catalog No. S07045


Add to Cart

Includes:
  • Dilute soap solution - 30mL
  • Hole punch
  • Plastic cups -16 count
  • Sodium bicarbonate - 50g
  • Syringes, 10mL - 16 count
  • Teacher's manual and student study guide

Requires :
  • Living plant leaves
  • Timers or stop watches
  • Light source
  • Distille or deionized water


Description

Description

Students will design and conduct an experiment using a selected variable to examine the rate of photosynthesis.

  • Meets AP Science Practices 1, 2, 3, 6 and 7, and Big Idea 2
  • Includes enough materials for eight lab groups
  • For grades 10-12

Specifications

Specifications

Meets AP science practices 1, 2, 3, 6 and 7 and big idea 2
White
13 to 20
30ml dilute soap solution, hole punch, 16 count plastic cups, 50g sodium bicarbonate, 10ml 16 count syringes, teacher's manual and student study guide
9.75 x 6.25 x 4 in.
AP Biology Investigation Kit #5: Photosynthesis
7 to 14
Learn the necessary components and conditions for photosynthesis to occur while using leaf disks to measure the accumulation of oxygen and relate it to the rate of photosynthesis
1
8 Lab Groups
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit