Learn More
Restek™ Injection Port Weldments
For Agilent GCs with OI Purge and Trap Systems
$651.00 - $659.00
Specifications
Type | GC |
---|---|
Injection Mode | Split/Splitless |
Requirements | Use with OI Purge and Traps |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
06-611-225
|
Restek™
22665 |
Agilent 6890 GCs |
Each for $651.00
|
|
|||||
06-611-229
|
Restek™
22669 |
Agilent 6890 GCs (With Large Canister Type Filter) |
Each for $659.00
|
|
|||||
Description
- Easily attach a purge and trap with pre-installed, low dead volume fittings
Specifications
GC | |
Split/Splitless | |
Use with OI Purge and Traps |