Learn More
TA Instruments INDIUM CALIBRATION MATERIAL
Supplier: TA Instruments 900902.901
Indium Calibration Material (not certified): standard indium calibration material (not certified) used for temperature calibration of DSC cells and TMA instruments.