Learn More
Sklar Instruments Sklar Steam indicator tape is used for visual assurance that the package has been exposed to the sterilizing process. The color of the strip changes from baige to black during the process. This tape is 3/4 inch X 60 yards. EA.
Supplier: Sklar Instruments 101131B
Sklar's Steam indicator tape is used for visual assurance that the package has been exposed to the sterilizing process. The color of the strip changes from baige to black during the process. This tape is 3/4 inch X 60 yards.