Learn More
CROSSTEX INTERNATIONAL 0.4 mL Mini Spore Ampules, 10^6, 50/pk + 5 negative controls
Supplier: CROSSTEX INTERNATIONAL OS15006
Crosstex Mini Spore Ampules are Self-Contained Biological Indicators for monitoring steam processes consisting of a hermetically sealed glass ampule which contains modified Trypticase Soy Broth (TSB) with a pH indicator and Geobacillus stearothermophilus with a population level of 10^6.