Learn More
CTL Scientific Supply Corp INDANTHRENE YELL PAP,200 STR
Supplier: CTL Scientific Supply Corp 90751
INDANTHRENE YELLOW PAPER - box of 200 strips (each strip 20 x 70 mm). Indanthrene yellow paper is used to determine the end point in the conversion of vat dyes to the leuco form.