Learn More
Vector Laboratories ImmPRESS-HRP Anti-Mouse IgG, Rat-Adsorbed (Peroxidase) Polymer Detection Kit, Made In Horse 15 ml
SDP
Supplier:  Vector Laboratories MP742215
Encompass_Preferred
This ImmPRESS-HRP Anti-Mouse IgG Rat-Adsorbed (Peroxidase) Polymer Detection Kit is made in horse and targets mouse primary antibodies. Rat adsorption minimizes interference from endogenous rat IgG when the system is used on rat tissue. The ImmPRESS reporter enzyme staining system uses novel conjugation chemistry to create a highly sensitive, ready-to-use, one-step, biotin-free detection system. A combination of highly active enzyme (peroxidase or alkaline phosphatase) and affinity-purified secondary antibodies results in crisp, strong staining. The IHC immunostaining protocol is simple. Following a blocking step with 2.5% horse serum, sections are incubated with primary antibody. After a brief wash, the appropriate ImmPRESS Reagent is added and incubated for 30 minutes. Sections are again rinsed and the slides are developed with a peroxidase or alkaline phosphatase substrate. The ImmPRESS Reagent is ready-to-use and stains 75-150 sections.
Catalog No. NC0013992