Learn More
Rockland Immunochemicals BLOTTO Immunoanalytical Grade (Non-Fat Dry Milk) - B501-0500
Supplier: Rockland Immunochemicals B5010500
Rockland Immunochemicals produces the following blocking buffer reagents: BSA, BLOTTO, BLOTTO A, BLOTTO B and Blocking Buffer for Fluorescent Western Blotting (see related products below). These products are intended to block non-specific binding of proteins in various immunological assays. Typically, western blotting and ELISA methods call for the addition of a blocking agent prior to the addition of primary antibody.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.