Learn More
Cole-Parmer Process Technology VS.1107 Immersion Heater, 316 SS, bayonet style, 100 watt
Supplier:  Cole-Parmer 3613500
Encompass
Single Element Heaters Encsed in 304 Stainless Steel or PTFE include a grounded solid-state heating element. Heaters are self-limiting for safer operation. Simple connection and compact design make these heaters ideal—even where space is limited. Heaers include 4-ft power cord with a three-prong plug; 240 VAC models do not include a plug.
Catalog No. NC0806880
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.