Learn More
Cole-Parmer IC-300-10DPC Basic Digital Immersion Circulator; 120 VAC
Supplier: Cole-Parmer 0126205
This versatile immersion circulator can be used in several ways. Use as a stand-alone water bath temperature controller, a pump for recirculating chilled liquid from an external chiller, or a pump for providing temperature control to an external device. The basic digital circulator digital LED display, ±0.01°C stability, overtemperature indicator light, pressure pump, and low-liquid level cutoff. Reach below ambient temperatures by using the cooling coil 12122-87 connected to an external chiller.†Requires external cooling coil, dip cooler, or flow cooler to get to temperatures below ambient.Please Note: Effective June 2022, the Techne High-Stability Digital Immersion Circulator Model number FTE10DPC has now been rebranded Cole-Parmer under Model number IC-300-10DPC. All the same great features and specifications, just with a new and improved look.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.