Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-33 Receptor Rat anti-Mouse, RB780, Clone: U29-93, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 755884
Description
IL-33 Receptor Monoclonal antibody specifically detects IL-33 Receptor in Mouse samples. It is validated for Flow CytometrySpecifications
IL-33 Receptor | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
U29-93 | |
RB780 | |
Il1rl1; ST2; ST2L; IL-33R alpha; IL-33Rα; IL33RA; Ly84; DER4; Fit-1 | |
Mouse IL-33R Recombinant Protein | |
RUO | |
17082 | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title