Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG2b, κ, Mouse, BV480, Clone: 27-35; Isotype Control, BD Horizon™
Mouse Monoclonal Isotype Control
Supplier: BD Biosciences 566080
Description
IgG2b, κ Monoclonal antibody specifically detects IgG2b, κ in samples. It is validated for Flow cytometry, Isotype control.Specifications
IgG2b κ | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Primary | |
IgG2b κ |
Flow Cytometry | |
27-35 | |
Brilliant Violet 480 | |
Anti-Dansyl; Isotype control (anti-Dansyl) | |
50 μg | |
Cell Biology | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction