Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1, κ Mouse, RB545, Isotype Control, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569284
This item is not returnable.
View return policy
Description
The X40 hybridoma was derived from the fusion of mouse Sp2/0-Ag 14 myeloma cells with spleen cells from BALB/c mice immunized with keyhole limpet hemocyanin (KLH). The hybridoma secretes monoclonal antibody that binds specifically to KLH, an antigen not expressed on human cells or human cell lines.Specifications
Flow Cytometry | |
X40 | |
RB545 | |
IgG1, kappa Isotype Control (Anti-KLH) | |
Keyhole limpet hemocyanin (KLH) | |
0.1 mg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Purified | |
Excitation Source: Blue 488 nm, Excitation Max: 496 nm, Emission Max: 545 nm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction