Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgG1, Kappa Mouse, APC, Isotype Control, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567155
This item is not returnable.
View return policy
Description
IgG1, Kappa Monoclonal antibody specifically detects IgG1, Kappa. It is validated for Flow Cytometry, Isotype controlSpecifications
igG1, Kappa | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
0.1 mg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Flow Cytometry | |
X40 | |
APC | |
IgG1, kappa Control (Anti-KLH) | |
Keyhole limpet hemocyanin (KLH) | |
RUO | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction