Learn More
Thermo Scientific IC Mixer Column [1 EA]
Supplier: Thermo Scientific 049135
gradient mixer with 2 MM column enabling precise eluent blending for gradient elution ion chromatography with low dead volume from inert wetted materials