Learn More
Sigma Aldrich Fine Chemicals Biosciences HypoThermosol FRS Preservation Solution, 100 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H4416100ML
Encompass
HypoThermosol is an optimized hypothermic (2-8°C) preservation media that enables improved and extended preservation of cells, tissues and organs. HypoThermosol-FRS is uniquely formulated to address the molecular-biological response of cells during the hypothermic preservation process.
Catalog No. NC1644381