Learn More
Cole-Parmer Oakton Economical Minidrum Hygrothermograph
Supplier: Cole-Parmer 0836970
This hygrothermograph is so compact it will fit almost anywhere; measures just 6.00''W x 7.38''H x 3.88''D. Hygrothermograph employs a quartz-controlled drive mechanism for constant speed; operates for 6 months on one AA battery (included). Chart rotation period is 7 days. Acrylic cover latches securely to protect paper and pens and is easily removable.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.