Learn More
Pickering Laboratories Inc HYDROLYSIS REAGENT 950ML 4/CS
Supplier: Pickering Laboratories Inc CB130.2
Hydrolysis Reagent for Carbamate analysis 0.075 M, case of 4 (950 mL /bottle) EPA 531.2