Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human RbAp48 (aa 1-14) Synthetic Peptide
Synthetic Peptide
Supplier: Invitrogen™ PEP084
Description
Human RbAp48 (residues 1-14) Synthetic Peptide, PEP-084, from Invitrogen.
PEP-084 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 1-14 of human RbAp48 with a carboxy-terminal cysteine added for carrier protein conjugation. The sequence of this peptide is (amino to carboxy terminus): M1-A-D-K-F-A-A-F-D-D-A-V-E-E14-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and endogenous RbAp48 catalog # PA1-869. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
5928 | |
purified | |
Human RbAp48 (aa 1-14) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q09028 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
RBBP4 | |
No preservative | |
50 μg | |
M1 - A - D - K - F - A - A - F - D - D - A - V - E - E14 - C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title