Learn More
STEMCELL Technologies Human Peripheral Blood Monocytes, Frozen, Size: 1 x 10^7 cells
Supplier: STEMCELL Technologies 70034
Peripheral blood (PB) primary human monocytes are isolated from mononuclear cells (MNCs) using the negative selection immunomagnetic technique. Monocytes are untouched and ready for any downstream application. PB was collected using the anticoagulant acid-citrate-dextrose solution A (ACDA).Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) on Primary Cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.