Learn More
HUMANCELLS BIOSCIENCES Human Normal Peripheral Blood Mononuclear Cells (PBMC) | Frozen | 10 million
Supplier: HUMANCELLS BIOSCIENCES PBMCC10M
Human Peripheral Blood Mononuclear Cells (PBMCs) are blood cells with a single round nucleus. These cells include lymphocytes (T, B and NK cells), monocytes and dendritic cells. PBMCs are parts of the immune system which are critical to cell-mediated and humoral immunity.