Learn More
Active Motif Human Negative Control Primer Set 2
Supplier: Active Motif 71002
Human Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with human samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple human cell lines.