Learn More
Science First™ Human Karyotype Kit
Study how karyotypes relate to certain genetic diseases.
Supplier: Science First™ 6900
Includes:
- Six karyotyping trays
- Six chromosome plates of each of the three types of cells
- Six plastic bags and labels
- Teacher's manual describing the experiment and providing background information
- Student copy-master of student guide for karyotyping process
Description
Kit enables high school biology students to see the differences in human chromosomes, especially some of the chromosomal abnormalities which may occur in the human population, such as Klinefelter's Syndrome and Turner's Syndrome.
- Cells arrested in metaphase are microscopically photographed and enlarged to a workable size to form a metaphase plate
- A karyotype is constructed by cutting out the photographed chromosomes and arranging them in a particular order for study
Specifications
Human Karyotype Kit | |
To study how karyotypes relate to certain genetic diseases | |
6 karyotyping trays, 6 chromosome plates of each of the three types of cells, 6 plastic bags and labels, teacher's manual, student copy-master of student guide for karyotyping process |