Learn More
Jackson Immuno Research Labs Human Gamma Globulin, 10mg
Supplier: Jackson Immuno Research Labs 009000002
Physical State: Sterile-filtered liquid. Storage: Store at 2-8°C under sterile conditions. Prepare working dilution on day of use. Expiration date: one year from date of receipt. The expiration date may be extended if test results are acceptable for the intended use.